1,3-Dibrom-5-(trifluormethoxy)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

343951-1G
SIAL343951-1GEA 88 EUR
SIAL343951-1G
1,3-Dibrom-5-(trifluormethoxy)benzol, Sigma-Aldrich®
1,3-Dibromo-5-(trifluoromethoxy)benzene
Formel: C₇H₃Br₂F₃O
MDL: MFCD00075280
CAS-Nummer: 207226-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR