4-Brom-2,3,5,6-tetrafluorbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

344052-5G
SIAL344052-5GEA 195 EUR
SIAL344052-5G
4-Brom-2,3,5,6-tetrafluorbenzonitril, Sigma-Aldrich®
4-Brom-2,3,5,6-tetrafluorbenzonitril
Formel: C₇BrF₄N
Molecular Weight: 253,9792 g/mol
MDL: MFCD00075282
CAS-Nummer: 17823-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR