Copper(II) gluconate ≥98%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate

M-Clarity™ Qualitätsstufe = MQ100

344419-100G
SIAL344419-100GEA 39.9 EUR
SIAL344419-100G
Copper(II) gluconate ≥98%, Sigma-Aldrich®
Copper(II) gluconate

Copper(II) D-gluconate powder in combination with silicon was used for the fabrication of carbon coated Si-Cu3Si composite as the anode materials for Li-ion batteries.

Formel: C₁₂H₂₂CuO₁₄
Molecular Weight: 453,84 g/mol
Schmelzpunkt: 155…157 °C (Lit.)
MDL: MFCD00075297
CAS-Nummer: 527-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR