Iron(II) gluconate hydrate ≥98%, Pulver, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

344427-25G
SIAL344427-25GEA 52.1 EUR
SIAL344427-25G
Iron(II) gluconate hydrate ≥98%, Pulver, Sigma-Aldrich®
Iron(II) gluconate hydrate
Formel: C₁₂H₂₄FeO₁₅
MDL: MFCD00150872
CAS-Nummer: 22830-45-1

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Tan
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Iron (Fe)- Titration by Na2S2O3 ≥11,3%
Optical Rotation c = 1%; Water 3,7 - 9,7°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR