4-Hydroxy-3-nitrobenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

344575-25G
SIAL344575-25GEA 298 EUR
SIAL344575-25G
4-Hydroxy-3-nitrobenzonitril, Sigma-Aldrich®
4-Hydroxy-3-nitrobenzonitril
Formel: C₇H₄N₂O₃
Molecular Weight: 164,12 g/mol
MDL: MFCD00070775
CAS-Nummer: 3272-08-0
EINECS: 221-899-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR