1,2-Dichlor-4-fluor-5-nitrobenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,5-Dichloro-2-fluoronitrobenzene

M-Clarity™ Qualitätsstufe = MQ100

345660-5G
SIAL345660-5GEA 60.8 EUR
SIAL345660-5G
1,2-Dichlor-4-fluor-5-nitrobenzol, Sigma-Aldrich®
1,2-Dichloro-4-fluoro-5-nitrobenzene
Formel: C₆H₂Cl₂FNO₂
Molecular Weight: 209,99 g/mol
MDL: MFCD00075330
CAS-Nummer: 2339-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR