2'-Fluor-6'-(trifluormethyl)acetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

345733-1G
SIAL345733-1GEA 40 EUR
SIAL345733-1G
2'-Fluor-6'-(trifluormethyl)acetophenon, Sigma-Aldrich®
2'-Fluor-6'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Molecular Weight: 206,14 g/mol
MDL: MFCD00061249
CAS-Nummer: 174013-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR