2-Chlor-4-hydroxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

346063-500MG
SIAL346063-500MGEA 95.5 EUR
SIAL346063-500MG
2-Chlor-4-hydroxybenzaldehyd, Sigma-Aldrich®
2-Chloro-4-hydroxybenzaldehyde
Formel: C₇H₅ClO₂
Molecular Weight: 156,57 g/mol
MDL: MFCD00052184
CAS-Nummer: 56962-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR