1-(Trifluoracetyl)imidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,2-Trifluoro-1-(1H-imidazol-1-yl)ethanone

346128-10G
SIAL346128-10GEA 84.2 EUR
SIAL346128-10G
1-(Trifluoracetyl)imidazol, Sigma-Aldrich®
1-(Trifluoracetyl)imidazol
Formel: C₅H₃F₃N₂O
Molecular Weight: 164.09 g/mol
MDL: MFCD00014501
CAS-Nummer: 1546-79-8
EINECS: 216-282-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR