2-Fluor-3-(trifluormethyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

346497-5G
SIAL346497-5GEA 68.7 EUR
SIAL346497-5G
2-Fluor-3-(trifluormethyl)benzaldehyd, Sigma-Aldrich®
2-Fluoro-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
MDL: MFCD00061246
CAS-Nummer: 112641-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR