5-Phenyl-1,3-cyclohexandion, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Phenylcyclohexane-1,3-dione

M-Clarity™ Qualitätsstufe = MQ200

346578-5G
SIAL346578-5GEA 40.6 EUR
SIAL346578-5G
5-Phenyl-1,3-cyclohexandion, Sigma-Aldrich®
5-Phenyl-1,3-cyclohexanedione
Formel: C₁₂H₁₂O₂
Molecular Weight: 188,23 g/mol
MDL: MFCD00051846
CAS-Nummer: 493-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR