3-(2-Chlorphenyl)propiononitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(2-Chlorophenyl)propanenitrile

346632-1G
SIAL346632-1GEA 39.6 EUR
SIAL346632-1G
3-(2-Chlorphenyl)propiononitril, Sigma-Aldrich®
3-(2-Chlorphenyl)propiononitril
Formel: C₉H₈ClN
Molecular Weight: 165,6200 g/mol
MDL: MFCD00019873
CAS-Nummer: 7315-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR