6-Amino-1-methyluracil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Amino-1-methylpyrimidine-2,4(1H,3H)-dione

M-Clarity™ Qualitätsstufe = MQ100

346799-25G
SIAL346799-25GEA 82.8 EUR
SIAL346799-25G
6-Amino-1-methyluracil, Sigma-Aldrich®
6-Amino-1-methyluracil
Formel: C₅H₇N₃O₂
Molecular Weight: 141,13 g/mol
MDL: MFCD00075366
CAS-Nummer: 2434-53-9
EINECS: 219-422-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR