Triethylsilylacetylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Triethylsilylethin

M-Clarity™ Qualitätsstufe = MQ100

346977-5G
SIAL346977-5GEA 49.9 EUR
SIAL346977-5G
Triethylsilylacetylen, Sigma-Aldrich®
Triethylsilylacetylen
Formel: C₈H₁₆Si
Molecular Weight: 140,3 g/mol
MDL: MFCD00075373
CAS-Nummer: 1777-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR