(Chlormethyl)dimethylphenylsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

347094-10G
SIAL347094-10GEA 157 EUR
SIAL347094-10G
(Chlormethyl)dimethylphenylsilan, Sigma-Aldrich®
(Chlormethyl)dimethylphenylsilan
Formel: C₉H₁₃ClSi
MDL: MFCD00040858
CAS-Nummer: 1833-51-8
EINECS: 217-392-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR