2-Chlor-3,5-difluorphenol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

347809-5G
SIAL347809-5GEA 404 EUR
SIAL347809-5G
2-Chlor-3,5-difluorphenol, Sigma-Aldrich®
2-Chlor-3,5-difluorphenol
Formel: C₆H₃ClF₂O
Molecular Weight: 164,54 g/mol
MDL: MFCD00075400
CAS-Nummer: 206986-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR