1,3-Dimethyluracil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dihydroxy-1,3-dimethylpyrimidin

M-Clarity™ Qualitätsstufe = MQ100

349801-5G
SIAL349801-5GEA 42.9 EUR
SIAL349801-5G
1,3-Dimethyluracil, Sigma-Aldrich®
1,3-Dimethyluracil
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
MDL: MFCD00038065
CAS-Nummer: 874-14-6
EINECS: 212-856-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR