Zinn(IV)chlorid 1 mol/l in Heptan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Zinntetrachlorid
M-Clarity™ Qualitätsstufe = MQ100
357499-100ML
SIAL357499-100MLEA
70.9
EUR
SIAL357499-100ML
Zinn(IV)chlorid 1 mol/l in Heptan, Sigma-Aldrich®
Zinn(IV)chlorid
Formel:
Cl₄Sn Molecular Weight: 260,52 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011242 CAS-Nummer: 7646-78-8 EINECS: 231-588-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...