7-Methoxy-2-benzofurancarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

357723-1G
SIAL357723-1GEA 42.1 EUR
SIAL357723-1G
7-Methoxy-2-benzofurancarbonsäure, Sigma-Aldrich®
7-Methoxybenzofuran-2-carboxylic acid
Formel: C₁₀H₈O₄
Molecular Weight: 192,17 g/mol
MDL: MFCD00060516
CAS-Nummer: 4790-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR