Methylthiosalicylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-mercaptobenzoate

M-Clarity™ Qualitätsstufe = MQ100

357758-25G 357758-5G
SIAL357758-25GEA 228 EUR
SIAL357758-25G SIAL357758-5G
Methylthiosalicylat, Sigma-Aldrich®
Methylthiosalicylat
Formel: C₈H₈O₂S
MDL: MFCD00060678
CAS-Nummer: 4892-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR