Benzo[b]thiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

357790-100G 357790-25G 357790-5G
SIAL357790-100GEA 268 EUR
SIAL357790-100G SIAL357790-25G SIAL357790-5G
Benzo[b]thiophen, Sigma-Aldrich®
Benzo[b]thiophen
Formel: C₈H₆S
MDL: MFCD00005864
CAS-Nummer: 95-15-8
EINECS: 202-395-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR