Mangan(II)perchlorat Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

359386-100G
SIAL359386-100GEA 86.3 EUR
SIAL359386-100G
Mangan(II)perchlorat Hydrat, Sigma-Aldrich®
Mangan(II)perchlorat Hydrat
Formel: Cl₂MnO₈·H₂O
CAS-Nummer: 698999-57-4
EINECS: 237-390-9
UN: 1481
ADR: 5.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR