Ammoniumdiethyldithiocarbamat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyldithiocarbamidsäure Ammoniumsalz, N,N-Diethyldithiocarbamidsäure Ammoniumsalz, (Diethyldithiocarbamato-S,S')ammonium

M-Clarity™ Qualitätsstufe = MQ200

359548-10G
SIAL359548-10GEA 135 EUR
SIAL359548-10G
Ammoniumdiethyldithiocarbamat, Sigma-Aldrich®
Ammoniumdiethyldithiocarbamat
Formel: C₅H₁₄N₂S₂
Molecular Weight: 166,31 g/mol
CAS-Nummer: 21124-33-4
EINECS: 244-224-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR