N(α)-Benzyloxycarbonyl-L-lysin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH

M-Clarity™ Qualitätsstufe = MQ200

359793-5G
SIAL359793-5GEA 107 EUR
SIAL359793-5G
N(α)-Benzyloxycarbonyl-L-lysin, Sigma-Aldrich®
N(α)-Benzyloxycarbonyl-L-lysin
Formel: C₁₄H₂₀N₂O₄
Molecular Weight: 280,3 g/mol
MDL: MFCD00038204
CAS-Nummer: 2212-75-1
EINECS: 218-662-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR