tert-Butylmethylmalonat, Sigma-Aldrich®

Synonyme: tert.-Butyl methyl malonate

M-Clarity™ Qualitätsstufe = MQ100

360015-10ML
SIAL360015-10MLEA 193 EUR
SIAL360015-10ML
tert-Butylmethylmalonat, Sigma-Aldrich®
tert-Butylmethylmalonat
Formel: C₈H₁₄O₄
Molecular Weight: 174.19 g/mol
MDL: MFCD00042856
CAS-Nummer: 42726-73-8
EINECS: 255-919-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR