trans-Bis(triphenylphosphin)palladium(II)bromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-Dibromobis(triphenylphosphine)palladium(II)

M-Clarity™ Qualitätsstufe = MQ100

360163-1G
SIAL360163-1GEA 95.5 EUR
SIAL360163-1G
trans-Bis(triphenylphosphin)palladium(II)bromid, Sigma-Aldrich®
trans-Bis(triphenylphosphin)palladium(II)bromid
Formel: C₃₆H₃₂Br₂P₂Pd
Molecular Weight: 792,81 g/mol
MDL: MFCD00134175
CAS-Nummer: 22180-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR