4,4'-Difluorbenzhydrylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chloro bis-(4-fluorophenyl)methane, 4,4-Difluorodiphenylmethylchloride

M-Clarity™ Qualitätsstufe = MQ100

360724-5G
SIAL360724-5GEA 36.5 EUR
SIAL360724-5G
4,4'-Difluorbenzhydrylchlorid, Sigma-Aldrich®
4,4'-Difluorbenzhydrylchlorid
Formel: C₁₃H₉ClF₂
Molecular Weight: 238,66 g/mol
MDL: MFCD00044329
CAS-Nummer: 27064-94-4
EINECS: 248-201-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR