4,4'-Methylenbis(2,6-diethylanilin), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(4-amino-3,5-diethylphenyl)methane, 4,4-Methylenebis(2,6-diethylaniline), 4,4'-Diamino-3,3',5,5'-tetraethyldiphenylmethane

360783-100G
SIAL360783-100GEA 127 EUR
SIAL360783-100G
4,4'-Methylenbis(2,6-diethylanilin), Sigma-Aldrich®
4,4'-Methylenbis(2,6-diethylanilin)
Formel: C₂₁H₃₀N₂
Molecular Weight: 310.4763 g/mol
MDL: MFCD00071552
CAS-Nummer: 13680-35-8
EINECS: 237-185-4
REACH: 01-0000015192-78

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR