4-Methoxy-2(5H)-furanon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

360872-5G
SIAL360872-5GEA 56.2 EUR
SIAL360872-5G
4-Methoxy-2(5H)-furanon, Sigma-Aldrich®
4-Methoxy-2(5H)-furanon
Formel: C₅H₆O₃
Molecular Weight: 114,1 g/mol
MDL: MFCD00071565
CAS-Nummer: 69556-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR