2,5-Dimethylstyrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

361135-5G
SIAL361135-5GEA 142 EUR
SIAL361135-5G
2,5-Dimethylstyrol, Sigma-Aldrich®
2,5-Dimethylstyrol
Formel: C₁₀H₁₂
Molecular Weight: 132,21 g/mol
MDL: MFCD00008614
CAS-Nummer: 2039-89-6
EINECS: 218-028-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR