1-(2-Methoxyphenyl)-2-thioharnstoff, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2-Methoxyphenylthioharnstoff, N-(2-Methoxyphenyl)thioharnstoff
361321-10G
SIAL361321-10GEA
91.6
EUR
SIAL361321-10G
1-(2-Methoxyphenyl)-2-thioharnstoff, Sigma-Aldrich®
1-(2-Methoxyphenyl)-2-thioharnstoff
Formel:
(C₆H₄(OCH₃))NC(S)NH₂ Molecular Weight: 182,25 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 156 °C |
MDL:
MFCD00022162 CAS-Nummer: 1516-37-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...