meso-2,3-Butandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: meso-2,3-Butylenglykol, (R,S)-2,3-Dihydroxybutan, (R,S)-2,3-Butylenglycol, (R,S)-2,3-Butandiol, (R,S)-2,3-Butylenglykol, meso-2,3-Butylenglycol, meso-2,3-Dihydroxybutan

M-Clarity™ Qualitätsstufe = MQ200

361461-10G
SIAL361461-10GEA 370 EUR
SIAL361461-10G
meso-2,3-Butandiol, Sigma-Aldrich®
meso-2,3-Butandiol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
MDL: MFCD00077669
CAS-Nummer: 5341-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR