Diethylaluminiumchlorid 1 mol/l in Heptan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DEAC

M-Clarity™ Qualitätsstufe = MQ100

361968-100ML
SIAL361968-100MLEA 81.2 EUR
SIAL361968-100ML
Diethylaluminiumchlorid 1 mol/l in Heptan, Sigma-Aldrich®
Diethylaluminiumchlorid
Formel: C₄H₁₀AlCl
Molecular Weight: 120,56 g/mol
MDL: MFCD00000459
CAS-Nummer: 96-10-6
EINECS: 202-477-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR