1,3-Dibenzoylpropan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,5-Diphenyl-1,5-butandion, 1,5-Diphenylbutan-1,5-dion

362069-5G
SIAL362069-5GEA 82.8 EUR
SIAL362069-5G
1,3-Dibenzoylpropan, Sigma-Aldrich®
1,3-Dibenzoylpropan
Formel: C₁₇H₁₆O₂
Molecular Weight: 252,31 g/mol
MDL: MFCD00051401
CAS-Nummer: 6263-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR