Methyl-trans-3-methoxyacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl trans-3-methoxyacrylate

M-Clarity™ Qualitätsstufe = MQ100

362379-100ML
SIAL362379-100MLEA 101 EUR
SIAL362379-100ML
Methyl-trans-3-methoxyacrylat, Sigma-Aldrich®
Methyl-trans-3-methoxyacrylat
Formel: C₅H₈O₃
Molecular Weight: 116,1152 g/mol
MDL: MFCD00128206
CAS-Nummer: 5788-17-0
EINECS: 412-900-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR