4-Chlor-2-fluor-5-nitrotoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

362417-5G
SIAL362417-5GEA 93.2 EUR
SIAL362417-5G
4-Chlor-2-fluor-5-nitrotoluol, Sigma-Aldrich®
4-Chlor-2-fluor-5-nitrotoluol
Formel: C₇H₅ClFNO₂
Molecular Weight: 189,57 g/mol
MDL: MFCD00134231
CAS-Nummer: 18349-11-6
EINECS: 242-225-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR