2-Fluor-5-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

362654-5G
SIAL362654-5GEA 22.3 EUR
SIAL362654-5G
2-Fluor-5-nitrobenzoesäure, Sigma-Aldrich®
2-Fluor-5-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00134238
CAS-Nummer: 7304-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR