Calcium-2-ethylhexanoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Ethylhexansäure Calciumsalz, Heptan-3-carbonsäure Calciumsalz, 2-Ethylcapronsäure Calciumsalz

362964-25G
SIAL362964-25GEA 200 EUR
SIAL362964-25G
Calcium-2-ethylhexanoat, Sigma-Aldrich®
Calcium-2-ethylhexanoat
Formel: C₁₆H₃₀CaO₄
Molecular Weight: 326,49 g/mol
MDL: MFCD00014001
CAS-Nummer: 136-51-6
EINECS: 205-249-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR