4-Chlorbutyrophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: γ-Chlorbutyrophenon, ω-Chlorbutyrophenon

363448-5G
SIAL363448-5GEA 63.6 EUR
SIAL363448-5G
4-Chlorbutyrophenon, Sigma-Aldrich®
4-Chlorbutyrophenon
Formel: (C₆H₅)C(O)(CH₂)₃Cl
Molecular Weight: 182,65 g/mol
MDL: MFCD00001005
CAS-Nummer: 939-52-6
EINECS: 213-362-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR