Benzyl-(R)-(-)-glycidylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-2-((Benzyloxy)methyl)oxirane

M-Clarity™ Qualitätsstufe = MQ100

363529-1G
SIAL363529-1GEA 35.2 EUR
SIAL363529-1G
Benzyl-(R)-(-)-glycidylether, Sigma-Aldrich®
Benzyl-(R)-(-)-glycidylether
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,20 g/mol
MDL: MFCD00068409
CAS-Nummer: 14618-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR