Benzyl-(S)-(+)-glycidylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2-((Benzyloxy)methyl)oxirane

M-Clarity™ Qualitätsstufe = MQ100

363537-1G
SIAL363537-1GEA 22.7 EUR
SIAL363537-1G
Benzyl-(S)-(+)-glycidylether, Sigma-Aldrich®
Benzyl-(S)-(+)-glycidylether
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,20 g/mol
MDL: MFCD00054428
CAS-Nummer: 16495-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR