2,3,5,6-Tetrafluor-4-hydroxybenzoesäure Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

363855-5G
SIAL363855-5GEA 162 EUR
SIAL363855-5G
2,3,5,6-Tetrafluor-4-hydroxybenzoesäure Hydrat, Sigma-Aldrich®
2,3,5,6-Tetrafluor-4-hydroxybenzoesäure Hydrat
Formel: C₇H₂F₄O₃·H₂O
Molecular Weight: 228,1 g/mol
CAS-Nummer: 207738-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR