(1S,2S)-(-)-1,2-Diphenylethylendiamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

364002-500MG
SIAL364002-500MGEA 111 EUR
SIAL364002-500MG
(1S,2S)-(-)-1,2-Diphenylethylendiamin, Sigma-Aldrich®
(1S,2S)-(-)-1,2-Diphenylethylendiamin
Formel: C₁₄H₁₆N₂
MDL: MFCD00082751
CAS-Nummer: 29841-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR