(1R,2R)-(+)-1,2-Diphenylethylendiamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

364010-1G 364010-500MG
SIAL364010-1GEA 94 EUR
SIAL364010-1G SIAL364010-500MG
(1R,2R)-(+)-1,2-Diphenylethylendiamin, Sigma-Aldrich®
(1R,2R)-(+)-1,2-Diphenylethylendiamin
Formel: C₁₄H₁₆N₂
MDL: MFCD00082769
CAS-Nummer: 35132-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR