Ethylmagnesiumbromid ∼13% (w/w) 1 mol/l in THF, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

364673-100ML 364673-800ML
SIAL364673-100MLEA 62.9 EUR
SIAL364673-100ML SIAL364673-800ML
Ethylmagnesiumbromid ∼13% (w/w) 1 mol/l in THF, Sigma-Aldrich®
Ethylmagnesium bromide
Formel: C₂H₅BrMg
MDL: MFCD00000043
CAS-Nummer: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR