Lithiummetaborat Dihydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

365297-500G
SIAL365297-500GEA 125 EUR
SIAL365297-500G
Lithiummetaborat Dihydrat, Sigma-Aldrich®
Lithiummetaborat Dihydrat
Formel: BLiO₂·2H₂O
Molecular Weight: 85,78 g/mol
MDL: MFCD00149768
CAS-Nummer: 15293-74-0
EINECS: 236-631-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR