4-Methyl-2-pyridon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-4-methylpyridin

365440-5G
SIAL365440-5GEA 83.7 EUR
SIAL365440-5G
4-Methyl-2-pyridon, Sigma-Aldrich®
4-Methyl-2-pyridon
Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
MDL: MFCD00134298
CAS-Nummer: 13466-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR