α'-Chlor-α,α,α-trifluor-p-xylol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-(Trifluormethyl)benzylchlorid, 4-(Chlormethyl)benzotrifluorid, 1-(Chlormethyl)-4-(trifluormethyl)benzol
M-Clarity™ Qualitätsstufe = MQ100
365815-5G
SIAL365815-5GEA
37.2
EUR
SIAL365815-5G
α'-Chlor-α,α,α-trifluor-p-xylol, Sigma-Aldrich®
α'-Chlor-α,α,α-trifluor-p-xylol
Formel:
C₈H₆ClF₃ Molecular Weight: 194,58 g/mol Siedepunkt: 47…49 °C (20 mmHg) Schmelzpunkt: 19 °C Dichte: 1,31 g/cm³ (20 °C) |
MDL:
MFCD00040772 CAS-Nummer: 939-99-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...