2,3-Dichlor-5-(trifluormethyl)pyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
366145-25G
SIAL366145-25GEA
39.6
EUR
SIAL366145-25G
2,3-Dichlor-5-(trifluormethyl)pyridin, Sigma-Aldrich®
2,3-Dichloro-5-(trifluoromethyl)pyridine
Formel:
C₆H₂Cl₂F₃N Molecular Weight: 215,99 g/mol Siedepunkt: 176 °C (1013 hPa) |
MDL:
MFCD00042243 CAS-Nummer: 69045-84-7 EINECS: 410-340-5 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...