Cyclopentancarbonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

366471-10G
SIAL366471-10GEA 288 EUR
SIAL366471-10G
Cyclopentancarbonitril, Sigma-Aldrich®
Cyclopentancarbonitril
Formel: C₆H₉N
Molecular Weight: 95,14 g/mol
MDL: MFCD00013741
CAS-Nummer: 4254-02-8
EINECS: 224-229-2
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR